Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146206.4 | NP_666318.2 | 23 - 24 | 2049 | 62 | |
GenBank mRNA | AK052930.1 | - | 23 - 24 | 2050 | 62 | |
AK089490.1 | - | 23 - 24 | 2113 | 62 | ||
AK170863.1 | - | 22 - 23 | 1995 | 62 | ||
BC025890.1 | - | 6 - 7 | 933 | 62 | ||
BC043472.1 | - | 19 - 20 | 1627 | 62 | ||
BC069857.1 | - | 6 - 7 | 737 | 62 | ||
BC141195.1 | - | 23 - 24 | 2133 | 9055807 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |