Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033318.3 | NP_001028490.2 | 2 - 3 | 423 | 119 | |
GenBank mRNA | AK030709.1 | - | 2 - 3 | 390 | 119 | |
AK145163.1 | - | 2 - 3 | 421 | 119 | ||
AK154693.1 | - | 2 - 3 | 425 | 119 | ||
AK163143.1 | - | 443 | 119 | |||
AK172200.1 | - | 2 - 3 | 425 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |