Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178381.3 | NP_848468.2 | 2 - 3 | 166 | 117 | |
GenBank mRNA | BC025477.1 | - | 2 - 3 | 134 | 117 | |
CF896275.1 | - | 2 - 3 | 166 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |