Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146205.2 | NP_666317.2 | 4 - 5 | 2627 | 79 | |
GenBank mRNA | AK030000.1 | - | 4 - 5 | 1484 | 79 | |
AK131111.1 | - | 4 - 5 | 2627 | 79 | ||
BC025196.1 | - | 1 - 2 | 98 | 79 | ||
BC032200.1 | - | 4 - 5 | 1972 | 5342166 | 79 | |
BC089363.1 | - | 4 - 5 | 1868 | 5720275 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |