Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029848.1 | NP_084124.1 | 2 - 3 | 167 | 132 | |
GenBank mRNA | AK014599.1 | - | 190 | 132 | ||
BC125402.1 | - | 2 - 3 | 167 | 40129917 | 132 | |
BC132096.1 | - | 2 - 3 | 167 | 40130373 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |