Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017478.3 | NP_059506.1 | 681 | 85 | ||
GenBank mRNA | AF205065.1 | - | 584 | 85 | ||
AJ251067.1 | - | 653 | 85 | |||
AK079599.1 | - | 8 - 9 | 597 | 85 | ||
AK088492.1 | - | 665 | 85 | |||
AK147943.1 | - | 627 | 85 | |||
AK149462.1 | - | 8 - 9 | 618 | 85 | ||
AK163668.1 | - | 8 - 9 | 652 | 85 | ||
BC049178.1 | - | 647 | 6309945 | 85 | ||
BC145898.1 | - | 624 | 40131201 | 85 | ||
BC145900.1 | - | 624 | 40131203 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |