Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Braf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139294.5 | NP_647455.3 | 1 - 2 | 360 | 74 | |
GenBank mRNA | AK140431.1 | - | 1 - 2 | 361 | 74 | |
BF466677.1 | - | 1 - 2 | 223 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Protein Kinase Mus musculus Probe spans exons |