Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146351.1 | NP_001139823.1 | 13 - 14 | 2772 | 77 | |
NM_007680.4 | NP_031706.3 | 13 - 14 | 2768 | 77 | ||
GenBank mRNA | AK076442.1 | - | 13 - 14 | 2595 | 77 | |
AK087423.1 | - | 13 - 14 | 2683 | 77 | ||
AK163823.1 | - | 4 - 5 | 490 | 77 | ||
BC031924.1 | - | 12 - 13 | 2380 | 4911959 | 77 | |
L77867.1 | - | 13 - 14 | 2462 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |