Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011302.3 | NP_035432.3 | 3 - 4 | 360 | 79 | |
GenBank mRNA | AF073780.1 | - | 3 - 4 | 223 | 79 | |
AF084561.1 | - | 3 - 4 | 231 | 79 | ||
AJ011381.1 | - | 3 - 4 | 211 | 79 | ||
BC046422.1 | - | 3 - 4 | 360 | 5401012 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |