Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176959.3 | NP_795933.2 | 1 - 2 | 550 | 73 | |
GenBank mRNA | AK048688.1 | - | 1 - 2 | 564 | 73 | |
AK051849.1 | - | 1 - 2 | 569 | 73 | ||
AK129227.1 | - | 1 - 2 | 498 | 73 | ||
BC050864.1 | - | 1 - 2 | 504 | 73 | ||
BC091646.1 | - | 1 - 2 | 550 | 30355382 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |