Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019635.2 | NP_062609.2 | 2 - 3 | 359 | 106 | |
GenBank mRNA | AF271361.1 | - | 2 - 3 | 105 | 106 | |
AY058922.1 | - | 2 - 3 | 245 | 106 | ||
BC049123.1 | - | 2 - 3 | 233 | 6813956 | 106 | |
U28726.1 | - | 2 - 3 | 238 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |