Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026449.3 | NP_080725.2 | 5 - 6 | 796 | 152 | |
GenBank mRNA | AK005605.1 | - | 5 - 6 | 817 | 152 | |
AK006193.1 | - | 5 - 6 | 775 | 152 | ||
BC049554.1 | - | 5 - 6 | 778 | 152 | ||
BC100447.1 | - | 5 - 6 | 649 | 152 | ||
BC145757.1 | - | 5 - 6 | 656 | 40131060 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |