Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011539.3 | NP_035669.3 | 8 - 9 | 989 | 56 | |
GenBank mRNA | AK048544.1 | - | 8 - 9 | 971 | 56 | |
AK085414.1 | - | 5 - 6 | 740 | 56 | ||
AK139479.1 | - | 8 - 9 | 952 | 56 | ||
BC029014.1 | - | 8 - 9 | 991 | 5007629 | 56 | |
L18868.1 | - | 8 - 9 | 860 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |