Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133925.2 | NP_598686.2 | 4 - 5 | 560 | 123 | |
GenBank mRNA | AK013047.1 | - | 4 - 5 | 530 | 123 | |
AK019296.1 | - | 4 - 5 | 509 | 123 | ||
AK156773.1 | - | 4 - 5 | 497 | 123 | ||
AK161670.1 | - | 4 - 5 | 525 | 123 | ||
AK167050.1 | - | 4 - 5 | 561 | 123 | ||
BC024602.1 | - | 4 - 5 | 441 | 123 | ||
BC040811.1 | - | 4 - 5 | 505 | 4015830 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |