Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011752.2 | NP_035882.1 | 12 - 13 | 1315 | 86 | |
GenBank mRNA | AB041612.1 | - | 12 - 13 | 1198 | 86 | |
AK016551.1 | - | 6 - 7 | 482 | 86 | ||
AK044472.1 | - | 12 - 13 | 1204 | 86 | ||
AK158995.1 | - | 12 - 13 | 1276 | 86 | ||
AK167849.1 | - | 12 - 13 | 1186 | 86 | ||
AK168061.1 | - | 12 - 13 | 1157 | 86 | ||
AK170372.1 | - | 3 - 4 | 1978 | 86 | ||
BC021397.1 | - | 12 - 13 | 1198 | 5067274 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |