Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281934.1 | NP_001268863.1 | 4 - 5 | 1465 | 87 | |
NM_008794.2 | NP_032820.2 | 4 - 5 | 1201 | 87 | ||
NR_104055.1 | 4 - 5 | 1201 | 87 | |||
GenBank mRNA | AK148416.1 | - | 4 - 5 | 786 | 87 | |
AK151980.1 | - | 4 - 5 | 788 | 87 | ||
AK165389.1 | - | 4 - 5 | 807 | 87 | ||
BC006730.1 | - | 3 - 4 | 702 | 3962441 | 87 | |
BC007322.1 | - | 4 - 5 | 1064 | 87 | ||
BC056456.1 | - | 4 - 5 | 1201 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |