Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021481.3 | NP_067456.1 | 5 - 6 | 539 | 103 | |
GenBank mRNA | AF136944.1 | - | 5 - 6 | 534 | 103 | |
AK008912.1 | - | 5 - 6 | 524 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |