Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016808.2 | NP_058088.2 | 5 - 6 | 1244 | 70 | |
NM_198091.2 | NP_932759.1 | 4 - 5 | 700 | 70 | ||
NM_198092.2 | NP_932760.2 | 5 - 6 | 1341 | 70 | ||
GenBank mRNA | AB041799.1 | - | 4 - 5 | 661 | 70 | |
AF079565.1 | - | 4 - 5 | 302 | 70 | ||
AK138799.1 | - | 6 - 7 | 1542 | 70 | ||
AY255637.1 | - | 4 - 5 | 669 | 70 | ||
AY255638.1 | - | 5 - 6 | 1223 | 70 | ||
AY255639.1 | - | 5 - 6 | 1177 | 70 | ||
AY255640.1 | - | 5 - 6 | 1222 | 70 | ||
BC017517.1 | - | 5 - 6 | 1223 | 4506362 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |