Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081412.2 | NP_001074881.1 | 3 - 4 | 1696 | 64 | |
GenBank mRNA | BC060270.1 | - | 3 - 4 | 934 | 64 | |
BC131682.1 | - | 4 - 5 | 1101 | 40111128 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |