Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177372.3 | NP_796346.2 | 3 - 4 | 505 | 69 | |
GenBank mRNA | AK028381.1 | - | 3 - 4 | 482 | 69 | |
AK129051.1 | - | 3 - 4 | 490 | 69 | ||
BC030382.1 | - | 3 - 4 | 447 | 69 | ||
BC115716.1 | - | 3 - 4 | 459 | 40085888 | 69 | |
CO433698.1 | - | 1 - 2 | 76 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |