Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030289.1 | NP_001025460.1 | 3 - 4 | 745 | 86 | |
GenBank mRNA | AK155813.1 | - | 4 - 5 | 732 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |