Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175535.3 | NP_780744.2 | 3 - 4 | 503 | 112 | |
GenBank mRNA | AK040756.1 | - | 3 - 4 | 503 | 112 | |
AK046916.1 | - | 3 - 4 | 510 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |