Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172924.3 | NP_766512.2 | 1 - 2 | 112 | 86 | |
GenBank mRNA | AK035440.1 | - | 1 - 2 | 102 | 86 | |
AK048912.1 | - | 1 - 2 | 101 | 86 | ||
AK090133.1 | - | 1 - 2 | 128 | 86 | ||
BC027695.1 | - | 3 - 4 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |