Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013900.2 | NP_038928.1 | 4 - 5 | 607 | 61 | |
GenBank mRNA | AB024336.1 | - | 4 - 5 | 607 | 61 | |
AK036444.1 | - | 4 - 5 | 542 | 61 | ||
BC040347.1 | - | 4 - 5 | 530 | 3672916 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |