Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178710.3 | NP_848825.2 | 5 - 6 | 801 | 77 | |
GenBank mRNA | AB067780.1 | - | 5 - 6 | 702 | 77 | |
AK090386.1 | - | 5 - 6 | 702 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |