Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024539.1 | NP_001019710.1 | 5 - 6 | 773 | 77 | |
GenBank mRNA | AK032878.1 | - | 6 - 7 | 458 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |