Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030261.4 | NP_084537.2 | 8 - 9 | 1494 | 94 | |
GenBank mRNA | AK017464.1 | - | 8 - 9 | 1391 | 94 | |
AK155398.1 | - | 8 - 9 | 1495 | 94 | ||
AK217306.1 | - | 1 - 2 | 88 | 94 | ||
BC132086.1 | - | 8 - 9 | 1315 | 40130363 | 94 | |
BC138690.1 | - | 8 - 9 | 1315 | 8861712 | 94 | |
KF543243.1 | - | 8 - 9 | 1243 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |