Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134007.4 | NP_598768.1 | 1 - 2 | 122 | 67 | |
GenBank mRNA | AK075907.1 | - | 1 - 2 | 123 | 67 | |
AK131595.1 | - | 1 - 2 | 123 | 67 | ||
AK131605.1 | - | 1 - 2 | 101 | 67 | ||
AK131812.1 | - | 1 - 2 | 126 | 67 | ||
AK131995.1 | - | 1 - 2 | 133 | 67 | ||
AK167898.1 | - | 1 - 2 | 101 | 67 | ||
BC013522.1 | - | 1 - 2 | 111 | 4236502 | 67 | |
BC019860.1 | - | 1 - 2 | 138 | 5123692 | 67 | |
BC021952.1 | - | 1 - 2 | 159 | 5102315 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |