Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012033.3 | NP_036163.3 | 6 - 7 | 978 | 59 | |
GenBank mRNA | AF153366.1 | - | 6 - 7 | 920 | 59 | |
AK008374.1 | - | 6 - 7 | 950 | 59 | ||
BC010745.1 | - | 6 - 7 | 923 | 4239192 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |