Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018758.2 | NP_061228.1 | 4 - 5 | 1019 | 86 | |
GenBank mRNA | AF070975.1 | - | 4 - 5 | 973 | 86 | |
AK045965.1 | - | 4 - 5 | 1020 | 86 | ||
AK082046.1 | - | 939 | 86 | |||
AK169462.1 | - | 4 - 5 | 1003 | 86 | ||
AK170182.1 | - | 4 - 5 | 967 | 86 | ||
BC005423.1 | - | 4 - 5 | 951 | 3487448 | 86 | |
BC009666.1 | - | 4 - 5 | 963 | 3490710 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |