Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnfn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081375.1 | NP_001074844.1 | 1 - 2 | 61 | 74 | |
GenBank mRNA | AK008979.1 | - | 1 - 2 | 62 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |