Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000019027.mm.1 |
NM_001163165.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019027.mm.1 |
NM_001163165.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019027.mm.1 |
NM_010440.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019027.mm.1 |
NM_010440.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019027.mm.1 |
TC1000002538.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019032.mm.1 |
NM_001163165.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019032.mm.1 |
NM_001163165.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019032.mm.1 |
NM_010440.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019032.mm.1 |
NM_010440.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019032.mm.1 |
TC1000002538.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019030.mm.1 |
NM_001163165.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019030.mm.1 |
NM_001163165.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019030.mm.1 |
NM_010440.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019030.mm.1 |
NM_010440.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019030.mm.1 |
TC1000002538.mm.1 |
MTA TC |