Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009719.6 | NP_033849.3 | 1 - 2 | 240 | 105 | |
GenBank mRNA | AK008017.1 | - | 1 - 2 | 241 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |