Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023219.2 | NP_075708.2 | 1 - 2 | 216 | 104 | |
GenBank mRNA | AF411960.2 | - | 1 - 2 | 136 | 104 | |
AK144296.1 | - | 159 | 104 | |||
BC109133.1 | - | 1 - 2 | 155 | 104 | ||
BC109134.1 | - | 1 - 2 | 155 | 104 | ||
BC127612.1 | - | 1 - 2 | 155 | 104 | ||
BC127615.1 | - | 1 - 2 | 155 | 40061657 | 104 | |
BC128465.1 | - | 1 - 2 | 155 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |