Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164376.1 | NP_001157848.1 | 11 - 12 | 1686 | 69 | |
NM_001164519.1 | NP_001157991.1 | 6 - 7 | 845 | 69 | ||
NM_177612.3 | NP_808280.2 | 11 - 12 | 1614 | 69 | ||
GenBank mRNA | AF344871.1 | - | 11 - 12 | 1686 | 69 | |
AK030166.1 | - | 11 - 12 | 1615 | 69 | ||
AK142745.1 | - | 11 - 12 | 1687 | 69 | ||
BC112409.1 | - | 11 - 12 | 1588 | 40060582 | 69 | |
DQ380429.1 | - | 11 - 12 | 1614 | 69 | ||
DQ380430.1 | - | 6 - 7 | 844 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |