Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033498.1 | NP_001028670.1 | 6 - 7 | 746 | 105 | |
GenBank mRNA | AK040545.1 | - | 7 - 8 | 635 | 105 | |
AK041631.1 | - | 6 - 7 | 746 | 105 | ||
BC138948.1 | - | 6 - 7 | 563 | 8861970 | 105 | |
BC138949.1 | - | 6 - 7 | 563 | 8861971 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |