Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201372.3 | NP_958760.2 | 8 - 9 | 730 | 72 | |
GenBank mRNA | AK052757.1 | - | 8 - 9 | 773 | 72 | |
BC055929.1 | - | 8 - 9 | 730 | 72 | ||
BC096560.1 | - | 8 - 9 | 735 | 1527981 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |