Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044389.2 | NP_001037854.1 | 7 - 8 | 1243 | 104 | |
NM_011802.3 | NP_035932.2 | 8 - 9 | 1285 | 104 | ||
GenBank mRNA | AF134983.1 | - | 8 - 9 | 1236 | 104 | |
AJ276991.1 | - | 8 - 9 | 1167 | 104 | ||
AK087848.1 | - | 8 - 9 | 1239 | 104 | ||
BC061153.1 | - | 7 - 8 | 1201 | 6593118 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |