Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175549.4 | NP_780758.3 | 16 - 17 | 2501 | 120 | |
GenBank mRNA | AK035110.1 | - | 17 - 18 | 3386 | 120 | |
AK085461.1 | - | 17 - 18 | 3137 | 120 | ||
AK129396.1 | - | 3 - 4 | 480 | 120 | ||
BC055333.1 | - | 17 - 18 | 2945 | 6409283 | 120 | |
DQ533875.1 | - | 18 - 19 | 2793 | 120 | ||
DQ533876.1 | - | 16 - 17 | 2501 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |