Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023655.2 | NP_076144.2 | 6 - 7 | 1641 | 94 | |
GenBank mRNA | AK013219.1 | - | 5 - 6 | 692 | 94 | |
AK028448.1 | - | 6 - 7 | 874 | 94 | ||
BC006699.1 | - | 6 - 7 | 1372 | 94 | ||
BC027353.1 | - | 6 - 7 | 1556 | 3988926 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |