Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025548.3 | NP_079824.2 | 2 | 556 | 61 | |
GenBank mRNA | AK002316.1 | - | 2 | 297 | 61 | |
AK003655.2 | - | 2 | 302 | 61 | ||
BC010684.1 | - | 2 | 289 | 2648112 | 61 | |
BQ445322.1 | - | 2 | 556 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |