Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028049.2 | NP_082325.2 | 4 - 5 | 578 | 82 | |
GenBank mRNA | AK002745.1 | - | 4 - 5 | 458 | 82 | |
AK005473.1 | - | 3 - 4 | 406 | 82 | ||
BC018273.1 | - | 4 - 5 | 518 | 4458296 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |