Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010149.3 | NP_034279.3 | 5 - 6 | 888 | 62 | |
GenBank mRNA | AK010968.1 | - | 4 - 5 | 596 | 62 | |
AK139031.1 | - | 5 - 6 | 787 | 62 | ||
BC003953.1 | - | 5 - 6 | 741 | 62 | ||
BC046282.1 | - | 5 - 6 | 818 | 6438591 | 62 | |
J04843.1 | - | 5 - 6 | 770 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |