Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
TC0700002788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
NM_019503.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
NM_019503.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
NM_052991.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
NM_052991.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
NM_052992.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
NM_052992.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
NM_194321.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024976.mm.1 |
NM_194321.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024980.mm.1 |
TC0700002788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024980.mm.1 |
NM_019503.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024980.mm.1 |
NM_019503.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024980.mm.1 |
NM_052992.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024980.mm.1 |
NM_052992.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
TC0700002788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
NM_019503.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
NM_019503.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
NM_052991.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
NM_052991.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
NM_052992.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
NM_052992.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
NM_194321.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024975.mm.1 |
NM_194321.2 |
Refseq |