Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010421.4 | NP_034551.2 | 4 - 5 | 514 | 100 | |
GenBank mRNA | AK075895.1 | - | 4 - 5 | 490 | 100 | |
AK075911.1 | - | 4 - 5 | 515 | 100 | ||
AK080777.1 | - | 4 - 5 | 487 | 100 | ||
AK144168.1 | - | 4 - 5 | 642 | 100 | ||
AK150444.1 | - | 4 - 5 | 485 | 100 | ||
AK151960.1 | - | 4 - 5 | 489 | 100 | ||
AK159091.1 | - | 4 - 5 | 506 | 100 | ||
AK159814.1 | - | 4 - 5 | 500 | 100 | ||
AK159890.1 | - | 4 - 5 | 487 | 100 | ||
AK160112.1 | - | 4 - 5 | 474 | 100 | ||
AK167013.1 | - | 4 - 5 | 500 | 100 | ||
AK168186.1 | - | 4 - 5 | 514 | 100 | ||
AK170903.1 | - | 4 - 5 | 500 | 100 | ||
BC010755.1 | - | 4 - 5 | 483 | 4225233 | 100 | |
U07631.1 | - | 4 - 5 | 506 | 100 | ||
X64331.1 | - | 4 - 5 | 457 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |