Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102455.1 | NP_001095925.1 | 15 - 16 | 2083 | 71 | |
NM_001102456.1 | NP_001095926.1 | 15 - 16 | 1951 | 71 | ||
NM_009691.2 | NP_033821.1 | 14 - 15 | 1915 | 71 | ||
GenBank mRNA | AK088391.1 | - | 2 - 3 | 306 | 71 | |
AK149977.1 | - | 15 - 16 | 2084 | 71 | ||
AK150020.1 | - | 15 - 16 | 2084 | 71 | ||
AK150478.1 | - | 7 - 8 | 815 | 71 | ||
AK151708.1 | - | 7 - 8 | 815 | 71 | ||
AK152787.1 | - | 7 - 8 | 815 | 71 | ||
AK153087.1 | - | 7 - 8 | 815 | 71 | ||
BC026491.1 | - | 5 - 6 | 367 | 71 | ||
BC051999.1 | - | 15 - 16 | 1929 | 5716148 | 71 | |
BC052396.1 | - | 14 - 15 | 1887 | 5719918 | 71 | |
M97216.1 | - | 10 - 11 | 1275 | 71 | ||
U15571.1 | - | 15 - 16 | 2059 | 71 | ||
Z22592.1 | - | 14 - 15 | 1915 | 71 | ||
Z27070.1 | - | 14 - 15 | 1913 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |