Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019459.2 | NP_062332.2 | 1 - 2 | 192 | 89 | |
GenBank mRNA | AF191090.1 | - | 1 - 2 | 520 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Probe spans exons |