Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028047.2 | NP_082323.1 | 2 - 3 | 426 | 87 | |
GenBank mRNA | AK005126.1 | - | 2 - 3 | 426 | 87 | |
AK039368.1 | - | 2 - 3 | 425 | 87 | ||
AK141576.1 | - | 2 - 3 | 417 | 87 | ||
AK161454.1 | - | 2 - 3 | 436 | 87 | ||
AK169651.1 | - | 2 - 3 | 350 | 87 | ||
BC034211.1 | - | 2 - 3 | 341 | 4988361 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |