Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010077.2 | NP_034207.2 | 3 - 4 | 485 | 102 | |
GenBank mRNA | BC105663.1 | - | 3 - 4 | 485 | 40046665 | 102 |
BC105664.1 | - | 3 - 4 | 485 | 40046668 | 102 | |
BC105665.1 | - | 3 - 4 | 485 | 40046660 | 102 | |
BC105666.1 | - | 3 - 4 | 485 | 40046662 | 102 | |
X55674.1 | - | 3 - 4 | 520 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |