Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199222.3 | NP_954692.2 | 2 - 3 | 436 | 84 | |
GenBank mRNA | AB211986.1 | - | 2 - 3 | 357 | 84 | |
BC020188.1 | - | 2 - 3 | 447 | 4925160 | 84 | |
BY708954.1 | - | 2 - 3 | 437 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |